Order Entry
Switzerland
ContactUsLinkComponent
2',7'-Dichlorofluorescin diacetate ≥94% (by HPLC), Calbiochem®
2',7'-Dichlorofluorescin diacetate ≥94% (by HPLC), Calbiochem®
Catalog # 287810-100
CAS Number:  
2',7'-Dichlorofluorescin diacetate ≥94% (by HPLC), Calbiochem®
Catalog # 287810-100
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥94%

Specification Test Results

About this item

 :
 :487.285 g/mol
 :Packaged under inert gas.
Storage Temperature:Freezer
CAS Number:4091-99-0
Formula:C₂₄H₁₆Cl₂O₇
MDL Number:MFCD00128955