Order Entry
Switzerland
ContactUsLinkComponent
PAD4 Autoantibody ELISA kit
PAD4 Autoantibody ELISA kit
Catalog # CAYM500930-96
Supplier:  Cayman Chemical
CAS Number:  
undefined
PAD4 Autoantibody ELISA kit
Catalog # CAYM500930-96
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    PAD4 Autoantibody ELISA kit, 96 wells
  • Detection range:
    15,6 - 1000 U/ml
  • Storage temperature:
    4 °C

Specifications

About this item

An immunometric for the measurement of anti-PAD4 autoantibodies.

  • Measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, IgA)
  • For use in human plasma and serum (no need to purify samples)
  • Lower Limit of quantification is 31,3 U/ml

PAD4 is a guanidino-modifying enzyme which catalyzes the conversion of specific arginine residues to citrulline in a calcium-dependent manner. PAD4 activity is increased in rheumatoid arthritis (RA), producing citrulline-containing proteins (CCP) that induce the formation of arthritogenic autoantibodies.

In addition to anti-CCP autoantibodies, 23 to 45% of RA patients also produce autoantibodies specific for the PAD4 enzyme itself. PAD4 Autoantibody ELISA Kit is an immunometric assay which can be used to measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, and IgA) in human plasma and serum without prior sample purification.

Affinity-purified anti-PAD4 antibody isolated from the plasma of a patient with RA is used as the standard. One unit is approximately equal to 1 ng of anti-PAD4 Ig protein.

Packaging: Kit contains: HRP conjugate, plate, ELISA standard, wash buffer concentrate, non-fat dry milk assay reagent, ELISA buffer concentrate, polysorbate 20, substrate and stop solution, cover sheet.