Water, meets analytical specification of Ph. Eur., USP for injection (WFI), cell culture reagent, sterile, Corning®
: Corning
: Hydrogen oxide
:7732-18-5
:
:H₂O
:MFCD00011332
:18.02 g/mol
:Ambient
- Environmentally Preferable:
- Mycoplasma:Tested Negative
- LM (plating effciency):Pass
- Oxidizable Substances:Pass
- Total Organic Carbon (before packaging) USP :<500 ppb
- Water Conductivity (packaged water) USP :Meets USP requirements
- Water Conductivity (before packaging) USP :Meets USP Temperature/Conductivity Requirements
- Endotoxin USP :≤0.025 EU/ml
- Sterility USP :Pass
- Acidity-Alkalinity:Tested in Accordance with EP
- Chlorides:Pass
- Nitrates:≤0.2 ppm
- Sulphates:Pass
- Ammonium:≤0.2 ppm
- Calcium and Magnesium:Pass
- Residue on Evaporation:≤3 mg (0.003)
Frequently Bought Together