Nalgene® Certified Platinum Clean HDPE Bottles and Carboys, Sterile
These certified platinum clean HDPE bottles and carboys help reduce the potential for carry-over contamination in biopharmaceutical and vaccine productions.
- Sterile and ready to use
- Washed with USP water
- Lot-to-lot tested
- Robust and versatile
These products include particulate certifications to under 1/3 the allowable limits according to USP <788> and lower endotoxin levels of 0,25 EU/ml, to help mitigate potential sources of unwanted impurities.
All container/closure systems certified leakproof to protect precious contents.
Can be used from –100 to 120 °C, allowing for both heated mixing and frozen storage or shipment. Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications.
: Particulate compliant and certified with Class 100 (ISO Class 5) cleanroom handling following washing and drying, including Class 10 (ISO Class 4) hoods for packaging.
: Products can be supplied with forced extraction studies, validation binders and certificates.
: Heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging.
- Catalog No:
- 216-2298
- 216-2299
- 216-2309
- 216-2304
- 216-2308
- 216-2307
- 216-2306
- 216-2300
- 216-2305
- 216-2303
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together