

Lithium perchlorate trihydrate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13453-78-6
:
Formula:ClLiO₄·3H₂O
MDL Number:MFCD00149765
:160.44 g/mol
Merck Index:14,05539
Density:1.84 g/cm³
Storage Temperature:Ambient
EINECS:232-237-2
UN:1481
ADR:5.1,II
:GHS03
:GHS07
Specification Test Results
- Assay (Unspecified):63.0 - 68.0% LiClO₄
- Heavy Metals (ICP-OES):5 ppm
- Impurity Content, Cl:0.003%
- Impurity Content, Fe:5 ppm
- Impurity Content, SO₄:0.001%
- Insoluble Material:0.005%
- pH:6.0 - 7.5
Specification Test Results
Related Information
Frequently Bought Together