Orders LinkContactUsLinkComponent
Latexometers
 :   Amarell
Latexometers
 :  Amarell
Latexometers
These instruments are calibrated in grams of dry rubber per litre of latex.
  • Without thermometer
  • Paper scale with well visible print
  • Shot weighted body
Accuracy: ±1%
Reference temperature: 28,9 °C
Product Family Options
  • Description
    Range
    Division
    Calibration temperature
    Length
  • Latexometer
    50 - 250 g/l
    10 g/l
    28,9 °C
    240 mm
    1 * 1 ST
    1 * 1 ST - SFr. 75.90
    SFr. 75.90
    1 * 1 ST
       SFr. 75.90
  • Latexometer
    50 - 450 g/l
    10 g/l
    28,9 °C
    210 mm
    1 * 1 ST
    1 * 1 ST - SFr. 75.90
    SFr. 75.90
    1 * 1 ST
       SFr. 75.90