ContactUsLinkComponent
EPA vials with PP cap
EPA vials with PP cap
  SCERSP60961C-4
undefined
EPA vials with PP cap
  SCERSP60961C-4
  • Cap included:
    Yes
  • Cap material:
    PP
  • Cap Size:
    24-400
  • Ø×H:
    28×60 mm
  • Septum:
    PTFE-faced silicone rubber
  • Capacity:
    20,00 ml
  • Colour:
    Clear
  • Description:
    EPA water analysis vials
  • Cap type:
    Screw cap, open-top
  • Cap colour:
    White
  • Pk:
    72
These clear screw thread EPA vials are used for the collection and storage of water samples.
  • Not pre-cleaned
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
 : These vials comply with the guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603 and 604).
 : Vials are packaged in a plastic shrink wrap module, lehr-clean, as they come from the end of the production lehr and are placed into a corrugated shelf-pack.