Descaler, Suma Calc D5
Katalognr. DIVYG11954
Supplier: Diversey
Specifications
- Capacity:10 l
- Description:Descaler, Suma Calc D5
- Packed:2× 5 litre
- Pk:2 Can
Specifications
About this item
This concentrated acidic descaler is mainly used to protect the equipment by improving the hygiene safety through effective removal of lime deposits.
- Lightening of dull metal surfaces by removal of oxide coatings
- Rapidly removes lime scale build up
- Corrosion inhibitor