Autoclave cleanroom labels, TX515A
Catalog # TEXWTX515A
Anbieter: Texwipe
Technische Daten
- Colour:White
- Description:Autoclavable labels, Teslin®
- Dimensions:38 mm
- Packed:500 labels/roll
- Pk:4 Roll
- Adhesive:Permanent acrylic
Technische Daten
Über diesen Artikel
These autoclavable labels are made of a Teslin® material designed for label printing and can be used with most conventional inks.
- Matte finish
- Permanent acrylic adhesive to provide excellent adhesion
- Prevent the removal of written data during the autoclave process
Ideal for applications requiring label and label data to maintain integrity through autoclave process.