VWR®, Colour reference solutions acc. to Ph. Eur., ready to use
Supplier: VWR Chemicals
Certificates
About this item
These solutions are developed and produced according to chapter 2.2.2. 'Degree of colouration of liquids' of the European Pharmacopoeia. (brown, brown-yellow, green-yellow, red and yellow). These kits are ready to use, and are dedicated to help you save time and money because preparation of these solutions is very complex.
- Easy to use kits
- Ready to use solutions
- Stable - shelf life of 36 months
- Certificate of Analysis provided with the kit
The kits are easy to use: Just add the sample into the empty vial provided with the kit and compare the colour with the reference solution in the rack.