Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Heating Cables, Moisture-Proof

Supplier:   HORST
Heating Cables, Moisture-Proof
Supplier:  HORST
Certificates
Heating Cables, Moisture-Proof

6 Options Available Below

Product Details & Documents

Moisture-proof heating cable with small diameter.

  • Thin and very flexible
  • Electrically insulated with PTFE
  • Bending radius of 10 mm

It is covered with a metallic protective braid, which is connected to the protective earth conductor. 1,5 m PTFE connection cable.

Nominal voltage of 230 V and temperature of 250 °C, with copper, nickel plated jacket.

Product Family Options

Product Information

  • Length
    Availability
    Price
  • HORS020821
    Heating cable Ø 2,5 - 3,5 mm, 30 W
    Heating cable Ø 2,5 - 3,5 mm, 30 W
    1,1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • HORS020822
    Heating cable Ø 2,5 - 3,5 mm, 50 W
    Heating cable Ø 2,5 - 3,5 mm, 50 W
    2 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • HORS020823
    Heating cable Ø 2,5 - 3,5 mm, 100 W
    Heating cable Ø 2,5 - 3,5 mm, 100 W
    4 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • HORS020824
    Heating cable Ø 2,5 - 3,5 mm, 120 W
    Heating cable Ø 2,5 - 3,5 mm, 120 W
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • HORS020803
    Heating cable Ø 2,5 - 3,5 mm, 200 W
    Heating cable Ø 2,5 - 3,5 mm, 200 W
    8 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • HORS020812
    Heating cable Ø 2,5 - 3,5 mm, 350 W
    Heating cable Ø 2,5 - 3,5 mm, 350 W
    14 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.