Orders LinkContactUsLinkComponent
Stand rods
Stand rods
Supplier:   USBECK Laborgeräte
Stand rods
Certificates
Stand rods
4 Options Available Below
Product Details & Documents
Steel, zinc plated.
  • Threaded (M 10)
  • Diameter: 12 mm
Product Family Options
Product Information
  • Ø
    Length
    Thread
    Availability
    Price
  • USBE2000/2100
    Steel rod, 400 g
    Steel rod, 400 g
    12 mm
    450 mm
    M 10
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 37.00
    SFr. 37.00
    1 * 1 ST
       SFr. 37.00
  • USBE2002/2102
    Steel rod, 530 g
    Steel rod, 530 g
    12 mm
    600 mm
    M 10
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 42.90
    SFr. 42.90
    1 * 1 ST
       SFr. 42.90
  • USBE2003/2103
    Steel rod, 650 g
    Steel rod, 650 g
    12 mm
    750 mm
    M 10
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 53.30
    SFr. 53.30
    1 * 1 ST
       SFr. 53.30
  • USBE2004/2104
    Steel rod, 880 g
    Steel rod, 880 g
    12 mm
    1000 mm
    M 10
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 70.80
    SFr. 70.80
    1 * 1 ST
       SFr. 70.80