ContactUsLinkComponent
Respirator, miniSCAPE
Respirator, miniSCAPE
Catalog # MSAA10038560
Supplier:  M S A
undefined
Respirator, miniSCAPE
Catalog # MSAA10038560
Supplier:  M S A
Specifications
  • Description:
    Respirator in plastic housing, miniSCAPE,
  • Pk:
    1
Specifications
About this item
Using cutting-edge technology, the miniSCAPE respirator provides minutes of safety when seconds count. The portable respirator uses the new-generation TabTec filter, fits in a user's pocket and protects against numerous gases and vapors.
  • Convenient, portable respirator small enough to fit in a user's pocket
  • For search and rescue purposes
Unit uses the TabTec filter to provide 5 minutes of protection and is housed in a rugged carrying case.
The miniSCAPE is fitted with a comfortable silicon mouthpiece directly connected to the filter housing. A nose clip is attached to the unit by a securing cord.
The miniSCAPE is stored in an impact resistant plastic case, which can be quickly and easily opened by pulling the sealed tab located on the front.
Certifications: DIN 58647-T7 (ABEK-5) duration: 5 minutes; CA 13.182.