Order Entry
Switzerland
ContactUsLinkComponent
Methyl orange (C.I. 13025), AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Methyl orange (C.I. 13025), AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Supplier:  VWR Chemicals
Certificates
undefined
Methyl orange (C.I. 13025), AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance of solution:
    Passes test
  • pH range (pink to orange-yellow):
    3,2 - 4,4
  • Visual transition interval:
    Passes test
  • Absorption maximum I1 (pH 3,1):
    501 - 504 nm
  • Absorption maximum I2 (pH 4,4):
    467 - 471 nm
  • Absorptivity (A1%, 1cm, I1, pH 3,1, on dried sample):
    1050 - 1150
  • Absorptivity (A1%, 1cm, I2, pH 4,4, on dried sample):
    750 - 850
  • Loss on drying (110 ºC):
    Max. 5%

Specification Test Results

Frequently Bought Together