Ethanol absolute (untaxed) ≥99.8%, Puriss. p.a. ACS, A15, without additive, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.790…0.791 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Refractive index (n 20/D):1.361 - 1.363
- Density (D 20/4):0.789 - 0.791
- Water (Karl Fischer):Max. 0.2 %
- Non-volatile matter:Max. 0.001 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.1 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 1 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Acetone (GC):Max. 10 ppm
- Acetone and 2-propanol according to ACS:complying
- Aldehydes (as CH₃CHO):Max. 10 ppm
- Formaldehyde:Max. 10 ppm
- APHA:Max. 10
- Refractive Index:n20/D 1.362
Specification Test Results
Frequently Bought Together