Order Entry
Switzerland
ContactUsLinkComponent
Celite® 545, filter aid ≥99% ≤125.30 μm (71% (acc. Cilas)), treated with sodium carbonate, flux calcined, Fluka™
Celite® 545, filter aid ≥99% ≤125.30 μm (71% (acc. Cilas)), treated with sodium carbonate, flux calcined, Fluka™
Catalog # 22140-5KG
CAS Number:  
Celite® 545, filter aid ≥99% ≤125.30 μm (71% (acc. Cilas)), treated with sodium carbonate, flux calcined, Fluka™
Catalog # 22140-5KG
CAS Number:  

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Composition:
    Al₂O₃, 1.0%
  • pH (10 % susp., 25°C):
    Min. 8.5
  • Loss on ignition (900°C):
    Max. 0.3 %

Specification Test Results

About this item

 :
 :60.08 g/mol
Storage Temperature:Ambient
EINECS:272-489-0
CAS Number:68855-54-9
Formula:SiO₂
MDL Number:MFCD00132803