Order Entry
Switzerland
ContactUsLinkComponent
Heptane (mixture of isomers) 99%, Reagent Grade
Heptane (mixture of isomers) 99%, Reagent Grade
Catalog # H2198-2.5L
CAS Number:  
Heptane (mixture of isomers) 99%, Reagent Grade
Catalog # H2198-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99 %
  • Refractive index (n 20/D):
    1.386 - 1.388
  • IR spectrum:
    complying
  • Appearance:
    complying
  • Vapor Pressure:
    40 mmHg ( 20 °C)
  • Vapor Density:
    3.5 (vs air)
  • Explosion Limit:
    7 %

Specification Test Results

About this item

 :
Merck Index:13,04679
Boiling Pt:6.7 %(V)
Melting Pt:–90.5 °C
Density:0.69 g/cm³ (15 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :100.2 g/mol
 :GHS02
 :GHS08
 :GHS07
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544