Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Hexane (mixture of isomers) ≥85% (isomers basis, GC), Puriss.
 
undefined
 
 
 
Hexane (mixture of isomers) ≥85% (isomers basis, GC), Puriss.

 

  • Assay of isomers (GC):
    Min. 85 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • Non-volatile matter:
    Max. 0.002 %
  • Aromatic substances (as C₆H₆) (GC):
    Max. 0.01 %
  • S-compounds (as S):
    Max. 0.005 %
  • n-Hexane (GC):
    Max. 4.9 %
  • Autoignition Temperature:
    437 °F
  • Vapor Density:
    3 (vs air)
  • Explosion Limit:
    7.5 %

 

Frequently Bought Together