Order Entry
Switzerland
ContactUsLinkComponent
Perchloric acid 0.01 M HClO4 (0.01N) in acetic acid volumetric solution, for titration in non-aqueous liquids, Fluka™
Corrosive
Flammable
Perchloric acid 0.01 M HClO4 (0.01N) in acetic acid volumetric solution, for titration in non-aqueous liquids, Fluka™
Catalog # 34408-1L
CAS Number:  
undefined
Corrosive
Flammable
Perchloric acid 0.01 M HClO4 (0.01N) in acetic acid volumetric solution, for titration in non-aqueous liquids, Fluka™
Catalog # 34408-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • c(perchloric acid):
    9.95 - 10.05 mM
  • Water (Karl Fischer):
    0.05 - 0.15 %
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:119 °C (1.013 hPa) (solvent)
Density:1.06 g/cm³ (20 °C)
Flash Pt:40 °C (The information regarding the flash point is that of the solvent)
Flash Pt:40 °C (solvent)
UN:2920
ADR:8,II
 :GHS05
 :GHS02
Storage Temperature:Ambient
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325