Order Entry
Switzerland
ContactUsLinkComponent
Calcium sulphate dihydrate 99.0-101.0% (by anhydrous basis), Puriss., meets analytical specification of NF, E516, Fluka™
Calcium sulphate dihydrate 99.0-101.0% (by anhydrous basis), Puriss., meets analytical specification of NF, E516, Fluka™
Certificates
Calcium sulphate dihydrate 99.0-101.0% (by anhydrous basis), Puriss., meets analytical specification of NF, E516, Fluka™

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Loss on drying (250°C):
    19.0 - 23.0 %
  • Arsenic (As):
    Max. 3 ppm
  • Iron (Fe):
    Max. 50 ppm
  • Mercury (Hg):
    Max. 1 ppm
  • Lead (Pb):
    Max. 2 ppm
  • Selenium (Se):
    Max. 30 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 0.01 %
  • Fluoride (F):
    Max. 30 ppm
  • Residual solvents:
    complying

Specification Test Results

Related Information

Frequently Bought Together