Order Entry
Switzerland
ContactUsLinkComponent
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Irritant
Health
Flammable
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Catalog # 33457-2.5L
CAS Number:  
Irritant
Health
Flammable
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Catalog # 33457-2.5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    123 - 126 °C
  • Explosion Limit:
    24.5 %
  • Density (D 20/20):
    0.966 - 0.968
  • Refractive index (n 20/D):
    1.4010 - 1.4030
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Titrable Acid:
    Max. 0.002 meq/g
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00004 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.0004 %
  • Acetaldehyde:
    Max. 0.001 %
  • Formaldehyde:
    Max. 0.001 %
  • Peroxides (as H₂O₂):
    Max. 0.0003 %
  • APHA:
    Max. 10

Specification Test Results

About this item

 :
 :76.1 g/mol
Merck Index:12,06118
Boiling Pt:124 °C (1.013 hPa)
Melting Pt:-85 °C
Melting Pt:–85 °C
Density:0.97 g/cm³ (20 °C)
Flash Pt:38 °C
EINECS:203-713-7
UN:1188
ADR:3,III
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:109-86-4
Formula:C₃H₈O₂
MDL Number:MFCD00002867