Order Entry
Switzerland
ContactUsLinkComponent
Manganese(II) sulphate monohydrate 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
Environment
Corrosive
Health
Manganese(II) sulphate monohydrate 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
Certificates
Environment
Corrosive
Health
Manganese(II) sulphate monohydrate 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™

Specification Test Results

  • Assay (calculated to ignited substance):
    99.0 - 100.5 %
  • Assay:
    98.0 - 102.0 %
  • Loss on ignition (500°C):
    10.0 - 12.0 %
  • Arsenic (As):
    Max. 3 ppm
  • Calcium (Ca):
    Max. 0.02 %
  • Iron (Fe):
    Max. 10 ppm
  • Magnesium (Mg):
    Max. 0.02 %
  • Lead (Pb):
    Max. 4 ppm
  • Selenium (Se):
    Max. 0.003 %
  • Zinc (Zn):
    Max. 50 ppm
  • Heavy metals (as Pb):
    Max. 0.002 %
  • Chloride (Cl):
    Max. 100 ppm
  • Substances not precipitated by (NH₄)2S:
    Max. 0.5 %
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

Specification Test Results

Related Information

Frequently Bought Together