Order Entry
Switzerland
ContactUsLinkComponent
Manganese(II) sulphate monohydrate 99.0-101.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
 
 
Manganese(II) sulphate monohydrate 99.0-101.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
  31425-50KG
 :  
 
 
Manganese(II) sulphate monohydrate 99.0-101.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
  31425-50KG
 :  

 

  • Pack type:
    Fiberboard box
  • Pk:
    50 kg
  • Seq:
    4418

 

 

  • assay:
    99.0 - 101.0 %
  • loss on ignition (400-500°C):
    10.0 - 12.0 %
  • water insoluble matter:
    Max. 0.005 %
  • calcium (Ca):
    Max. 0.005 %
  • copper (Cu):
    Max. 0.0005 %
  • iron (Fe):
    Max. 0.001 %
  • potassium (K):
    Max. 0.005 %
  • magnesium (Mg):
    Max. 0.005 %
  • sodium (Na):
    Max. 0.005 %
  • nickel (Ni):
    Max. 0.0005 %
  • lead (Pb):
    Max. 0.001 %
  • zinc (Zn):
    Max. 0.001 %
  • heavy metals (ICP) ACS:
    Max. 0.002 %
  • chloride (Cl):
    Max. 0.001 %
  • KMnO4 red. matter (as O):
    Max. 0.0005 %
  • solubility:
    complying
  • appearance of the solution:
    complying

 

 

 :
 :13,05763
 :Decomposes on heating
 : Dec. (on heating)
 :700 °C (anhydr.)
 :2.950 g/cm³ (20 °C)
 :Ambient
 :232-089-9
 :9,III
 :169.02 g/mol
 :GHS05
 :GHS08
 :10034-96-5
 :MnSO₄·1H₂O
 :MFCD00149159