Ammonium iron(II) sulphate hexahydrate ≥99.0% (RT), Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7783-85-9
:
Formula:(NH₄)₂Fe(SO₄)₂·6H₂O
MDL Number:MFCD00150530
:392.14 g/mol
Melting Pt:100 °C (dec.)
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-151-8
Specification Test Results
- Assay (manganometric):Min. 99.0 %
- pH (5 %, 25 °C):3.0 - 5.0
- Calcium (Ca):Max. 20 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 20 ppm
- Chromium (Cr):Max. 50 ppm
- Copper (Cu):Max. 20 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 100 ppm
- Manganese (Mn):Max. 500 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 200 ppm
- Lead (Pb):Max. 10 ppm
- Zinc (Zn):Max. 20 ppm
- Iron(III):Max. 200 ppm
- Chloride (Cl):Max. 10 ppm
- Phosphate (PO₄):Max. 20 ppm
Specification Test Results
Frequently Bought Together