Order Entry
Switzerland
ContactUsLinkComponent
 
(±)-α-Methoxyphenylacetic acid ≥99.0% (by titrimetric analysis) for gravimetric determination of Na, Fluka™
 
(±)-α-Methoxyphenylacetic acid ≥99.0% (by titrimetric analysis) for gravimetric determination of Na, Fluka™

 

  • Identity (NMR):
    complying
  • Assay:
    99.0 - 101.1 %
  • Residue on ignition:
    Max. 0.05 %
  • Melting point:
    69 - 71 °C
  • APHA:
    Max. 50

 

 

Frequently Bought Together