Ammonium hydrogen carbonate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., E503, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Ammonium bicarbonate
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
Merck Index:13,00497
Melting Pt:60 °C
Density:1.58 g/cm³ (20 °C)
EINECS:213-911-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Non-volatile matter:Max. 100 ppm
- Arsenic (As):Max. 1 ppm
- Iron (Fe):Max. 5 ppm
- Mercury (Hg):Max. 1 ppm
- Lead (Pb):Max. 2 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Chloride (Cl):Max. 10 ppm
- Sulfate (SO₄):Max. 30 ppm
- Residual solvents:complying
Specification Test Results
Frequently Bought Together