Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
Health
Flammable
Pyridine ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 270407-1L
CAS Number:  
undefined
Irritant
Health
Flammable
Pyridine ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 270407-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0004 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 350 nm:
    Max. 0.01
  • Absorbance at 335 nm:
    Max. 0.02
  • Absorbance at 315 nm:
    Max. 0.15
  • Absorbance at 305 nm:
    Max. 1.0
  • Autoignition Temperature:
    899 °F
  • Explosion Limit:
    12.4 %
  • Refractive Index:
    n20/D 1.509 (lit.)
  • Vapor Pressure:
    10 mmHg ( 13.2 °C)

Specification Test Results

About this item

 :
Merck Index:13,08060
Boiling Pt:115 °C (1.013 hPa)
Melting Pt:-42 °C
Melting Pt:–42 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:12.4 %(V)
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
 :79.1 g/mol
 :GHS07
 :GHS08
 :GHS02
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732