Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Silver nitrate ≥99.0% ACS, Fluka™
 
undefined
 
 
Silver nitrate ≥99.0% ACS, Fluka™

 

  • Assay:
    Min. 99.0 %
  • Free acid:
    complying
  • Copper (Cu):
    Max. 0.0002 %
  • Iron (Fe):
    Max. 0.0002 %
  • Lead (Pb):
    Max. 0.001 %
  • Substances not precipitated by HCl (as SO₄):
    Max. 0.01 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 0.002 %
  • Appearance of the solution:
    complying

 

 

Frequently Bought Together