
Sulphuric acid 95-97%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:310 °C (1.013 hPa)
Melting Pt:–14…–10 °C
Density:1.840 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
:GHS05
Specification Test Results
- Assay:95.0 - 97.0 %
- Residue on ignition:Max. 0.0005 %
- Arsenic (As):Max. 0.1 ppm
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0005 %
- Cobalt (Co):Max. 0.0005 %
- Chromium (Cr):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 2 ppm
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.0005 %
- Manganese (Mn):Max. 0.0005 %
- Sodium (Na):Max. 0.005 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 1 ppm
- Nitrate (NO₃):complying
- Appearance of the solution:complying
- Residual solvents:complying
Specification Test Results
Frequently Bought Together