Order Entry
Switzerland
ContactUsLinkComponent
Sodium sulphate, anhydrous 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Sodium sulphate, anhydrous 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Certificates
undefined
Sodium sulphate, anhydrous 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 100.5 %
  • Loss on drying (130°C):
    Max. 0.5 %
  • Free acid (as H₂SO₄):
    Max. 0.05 %
  • Free alkali (as NaOH):
    Max. 0.04 %
  • Arsenic (As):
    Max. 0.0002 %
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Zinc (Zn):
    Max. 0.03 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.02 %
  • Appearance of the solution:
    complying
  • Acidity or alkalinity:
    complying
  • Residual solvents:
    complying

Specification Test Results

Related Information

Frequently Bought Together