ContactUsLinkComponent
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Corrosive
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide 1 M NaOH (1N) ≥99.5% volumetric solution, Fluka™
Certificates
Specification Test Results
  • Calibrated Against:
    potassium hydrogen phthalate
  • Assay:
    0.998 - 1.002 mol/l