

Sodium iodide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1.304 °C (1.013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-679-3
ADR:9,III
:GHS07
:GHS08
Specification Test Results
- Assay (calculated to the dried substance):99.0 - 100.5 %
- Loss on drying (105°C):Max. 3.0 %
- Water (Karl Fischer):Max. 2.0 %
- Alkalinely reacting impurities:complying
- Iron (Fe):Max. 5 ppm
- Potassium (K):complying
- Heavy metals (as Pb):Max. 5 ppm
- Iodate (IO₃):Max. 2 ppm
- Iodate (JO₃):complying
- Sulfate (SO₄):Max. 150 ppm
- Thiosulfate (S₂O₃):Max. 0.005 %
- Thiosulfate and Barium:complying
- Nitrate, nitrite, ammonium:complying
- Residual solvents:complying
- Appearance of the solution:complying
Specification Test Results
Frequently Bought Together