Potassium bromide for IR spectrophotometry, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7758-02-3
:
Formula:KBr
MDL Number:MFCD00011358
Merck Index:13,07701
Boiling Pt:1.383 °C (1.013 hPa)
Melting Pt:734 °C
Density:2.73 g/cm³ (20 °C)
:119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
Specification Test Results
- Suitability for IR spectroscopy:complying
- Vapor Pressure:<0.01 mmHg ( 20 °C)
Specification Test Results
Frequently Bought Together