Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Tetrachloroethylene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
  270393-1L
 :  
undefined
 
 
Tetrachloroethylene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
  270393-1L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.90 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Absorbance at 400 nm:
    Max. 0.03
  • Absorbance at 350 nm:
    Max. 0.05
  • Absorbance at 305 nm:
    Max. 0.10
  • Absorbance at 300 nm:
    Max. 0.20
  • Absorbance at 295 nm:
    Max. 0.30
  • Absorbance at 290 nm:
    Max. 1.0
  • APHA:
    Max. 10
  • Vapor Pressure:
    13 mmHg ( 20 °C)
  • Vapor Density:
    5.83 (vs air)
  • Refractive Index:
    n20/D 1.505 (lit.)

 

 

 :
 :165.83 g/mol
 :12,09332
 :120-122 °C (1.013 hPa)
 :120…122 °C (1.013 hPa)
 :-22 °C
 :–22 °C
 :1.622…1.624 g/cm³ (20 °C)
 :Ambient
 :204-825-9
 :1897
 :6.1,III
 :GHS07
 :GHS08
 :127-18-4
 :C₂Cl₄
 :MFCD00000834