


n-Pentane ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:36…37 °C
Melting Pt:–131 °C
Density:0.631 g/cm³ (15 °C)
Flash Pt:<–35 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.357 - 1.359
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.05 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Isopentane (GC):Max. 1.0 %
- 2-methylpentane (GC):Max. 1.0 %
Specification Test Results
Frequently Bought Together