

Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
:60-29-7
:
:(CH₃CH₂)₂O
:MFCD00011646
:74.12 g/mol
:12,03852
:34…35 °C (1.013 hPa)
:–116 °C (1.013 hPa)
:0.71 g/cm³ (20 °C)
:–45 °C (1.013 hPa)
:Ambient
:1155
:3,I
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:200-467-2
:01-2119535785-29
:GHS02
:GHS07
- Appearance (color):complying
- Appearance (form):complying
- Assay (GC):Min. 99.8 %
- Boiling range:34 - 35 °C
- Density (D 20/4):0.713 - 0.714
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.03 %
- Free acid (as CH₃COOH):Max. 0.0002 %
- Titrable Acid:Max. 0.0002 meq/g
- Aluminium (Al):Max. 0.50 ppm
- Boron (B):Max. 0.02 ppm
- Barium (Ba):Max. 0.10 ppm
- Calcium (Ca):Max. 0.50 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.05 ppm
- Magnesium (Mg):Max. 0.10 ppm
- Manganese (Mn):Max. 0.02 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.05 ppm
- Tin (Sn):Max. 0.10 ppm
- Zinc (Zn):Max. 0.05 ppm
- Acetone (GC):Max. 0.005 %
- Aldehydes:complying
- Carbonyl compounds (as CO):Max. 0.001 %
- Carbonyl Compounds (as HCHO):Max. 0.001 %
- Ethanol (GC):Max. 0.02 %
- Methanol (GC):Max. 0.02 %
- Peroxides (as H₂O₂):Max. 0.3 ppm
- Reaction against H₂SO₄:complying
- APHA:Max. 10
- Assay BHT (GC):5 - 8 ppm
Frequently Bought Together