Order Entry
Switzerland
ContactUsLinkComponent
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Certificates
undefined
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • Titrable Acid:
    Max. 0.0002 meq/g
  • Peroxides (as H₂O₂):
    Max. 1 ppm
  • Ethanol (GC):
    Max. 0.05 %
  • Carbonyl Compounds (as HCHO):
    Max. 0.001 %
  • Assay BHT (GC):
    Min. 1 ppm
  • APHA:
    Max. 10
  • Vapor Pressure:
    8.56 psi ( 20 °C)
  • Refractive Index:
    n20/D 1.3530 (lit.)

Specification Test Results

Related Information

Frequently Bought Together