Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Dichloromethane ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
  650463-1L
 :  
undefined
 
 
Dichloromethane ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
  650463-1L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance (color):
    complying
  • Appearance (form):
    complying
  • Assay (GC):
    Min. 99.9 %
  • IR spectrum:
    complying
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.02
  • Absorbance at 240 nm:
    Max. 0.10
  • Absorbance at 235 nm:
    Max. 1.00
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb
  • 2-methyl-2-buten (GC):
    50 - 150 ppm
  • Suitability for residue analysis:
    Max. 10 ng/l
  • Vapor Pressure:
    24.45 psi ( 55 °C)

 

 

 :
 :84.93 g/mol
 :12,06140
 :39…41 °C (1.013 hPa)
 :–97 °C
 :1.32…1.33 g/cm³ (20 °C)
 :Ambient
 :200-838-9
 :1593
 :6.1,III
 :Stabilised with amylene 50-150 ppm
 :GHS07
 :GHS08
 :75-09-2
 :CH₂Cl₂
 :MFCD00000881