

Acetic acid ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS05
:GHS02
Specification Test Results
- Assay:99.8 - 100.5 %
- Assay (GC):Min. 99.8 %
- Density (D 20/20):1.052 - 1.053
- Boiling range:117 - 119 °C
- Congealing - Freezing Point:Min. 16.3 °C
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.4 %
- Silver (Ag):Max. 0.000001 %
- Aluminium (Al):Max. 0.000005 %
- Arsenic (As):Max. 0.000001 %
- Barium (Ba):Max. 0.000001 %
- Beryllium (Be):Max. 0.000001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00002 %
- Cadmium (Cd):Max. 0.000002 %
- Cobalt (Co):Max. 0.000001 %
- Chromium (Cr):Max. 0.000005 %
- Copper (Cu):Max. 0.000001 %
- Iron (Fe):Max. 0.00002 %
- Germanium (Ge):Max. 0.000005 %
- Potassium (K):Max. 0.00001 %
- Lithium (Li):Max. 0.000001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.000002 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000005 %
- Lead (Pb):Max. 0.000002 %
- Strontium (Sr):Max. 0.000001 %
- Titanium (Ti):Max. 0.000001 %
- Thallium (Tl):Max. 0.000005 %
- Vanadium (V):Max. 0.000001 %
- Zinc (Zn):Max. 0.000005 %
- Zirconium (Zr):Max. 0.00001 %
- Heavy metals (as Pb):Max. 0.00005 %
- Chloride (Cl):Max. 0.00005 %
- Phosphate (PO₄):Max. 0.00005 %
- Sulfate (SO₄):Max. 0.00005 %
- Acetaldehyde:Max. 0.0002 %
- Acetic aNHydride (GC):Max. 0.01 %
- Mixable with H₂O:complying
- Titrable Base:Max. 0.0004 meq/g
- APHA:Max. 10
- Appearance of the substance:complying
- K₂Cr₂O₇ reducing matter:complying
- KMnO₄ reducing substances:complying
Specification Test Results
Frequently Bought Together