

Ethyl acetate ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Merck Index:13,03792
Boiling Pt:77 °C (1.013 hPa)
Melting Pt:–84 °C
Density:0.901 g/cm³
Flash Pt:11.5 %(V)
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.7 %
- Non-volatile matter:max. 0.0005 %
- Water (Karl Fischer):max. 0.03 %
- Free acid (as CH₃COOH):Max. 0.005 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 300 nm:Max. 0.01
- Absorbance at 270 nm:Max. 0.10
- Absorbance at 260 nm:Max. 0.30
- Absorbance at 255 nm:Max. 0.70
- Refractive Index:n20/D 1.3720 (lit.)
Specification Test Results
Frequently Bought Together