Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
    
 
undefined
 
 
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Autoignition Temperature:
    801 °F
  • Assay (GC):
    Min. 99.9 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Vapor Density:
    3 (20 °C, vs air)
  • Vapor Pressure:
    73 mmHg ( 20 °C)

 

 

Frequently Bought Together