Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Ethyl acetate ≥99.8%, Biotechnology Grade, Burdick & Jackson™
    
 
undefined
 
 
Ethyl acetate ≥99.8%, Biotechnology Grade, Burdick & Jackson™
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Titrable Acid:
    Max. 0.0009 meq/g
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 275 nm:
    Max. 0.01
  • Absorbance at 263 nm:
    Max. 0.05
  • Absorbance at 254 nm:
    Max. 1.00
  • Vapor Pressure:
    73 mmHg ( 20 °C)

 

 

Frequently Bought Together