

Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:55…57 °C (1.013 hPa)
Melting Pt:–95 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:14.3 %(V)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Boiling range:55.5 - 56.5 °C
- Density (D 20/4):0.790 - 0.792
- Refractive index (n 20/D):1.3580 - 1.3600
- Non-volatile matter:Max. 10 ppm
- Water insoluble substances:complying
- Water (Karl Fischer):Max. 0.2 %
- Free acid (as CH₃COOH):Max. 0.0015 %
- Free alkali (as NH₃):Max. 0.0008 %
- Acidity or alkalinity:complying
- Aluminium (Al):Max. 0.5 ppm
- Boron (B):Max. 0.02 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.02 ppm
- Tin (Sn):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.05 ppm
- KMnO₄ reducing matter (as O):Max. 0.0002 %
- Reducing impurities:complying
- Aldehydes (as HCHO):Max. 0.001 %
- Benzene (GC):Max. 2 Vol.ppm
- Ethanol (GC):Max. 0.01 %
- Methanol (GC):Max. 0.05 Vol.%
- 2-propanol (GC):Max. 0.05 Vol.%
- Related substances (GC):complying
- Appearance of the solution:complying
- APHA:Max. 10
- Solubility in water:complying
Specification Test Results
Frequently Bought Together