Order Entry
Switzerland
ContactUsLinkComponent
Formaldehyde 37% (w/w) in water stabilised
Toxic
Health
Corrosive
Formaldehyde 37% (w/w) in water stabilised
Certificates
Toxic
Health
Corrosive
Formaldehyde 37% (w/w) in water stabilised
Certificates

Specification Test Results

  • Vapor Density:
    1.03 (vs air)
  • Assay:
    37 - 38 %
  • Free acid (as HCOOH):
    Max. 0.03 %
  • Methanol (GC):
    10 - 15 Vol.%
  • APHA:
    Max. 10
  • Autoignition Temperature:
    572 °F

Specification Test Results

Related Information

Frequently Bought Together