Order Entry
Switzerland
ContactUsLinkComponent
Lead(II) acetate trihydrate ≥99.5%, puriss., extra pure, Fluka™
Health
Environment
Lead(II) acetate trihydrate ≥99.5%, puriss., extra pure, Fluka™
Catalog # HONC11504-25KG
CAS Number:  
Health
Environment
Lead(II) acetate trihydrate ≥99.5%, puriss., extra pure, Fluka™
Catalog # HONC11504-25KG
CAS Number:  

Specifications

  • Pack type:
    Paper bag
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.001 %
  • Substances not precipitated by H₂S (SO₄):
    Max. 0.5 %
  • Chloride (Cl):
    Max. 0.002 %

Specification Test Results

About this item

 :
 :379.34 g/mol
Merck Index:13,05415
Boiling Pt:280 °C (1013 hPa)
Melting Pt:75 °C
Density:2.55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-104-4
UN:1616
ADR:6.1,III
 :GHS08
 :GHS09
CAS Number:6080-56-4
Formula:(H₃CCOO)₂Pb·3H₂O
MDL Number:MFCD00150023