Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis) ACS, Riedel-de Haën™
 
undefined
 
 
 
Xylene (mixture of isomers) ≥98.5% (xylenes + ethylbenzene basis) ACS, Riedel-de Haën™
 

 

  • Assay Xylene isomers and Ethylbenzol (GC):
    Min. 98.50 %
  • Ethyl benzene (GC):
    Max. 25 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Non-volatile matter:
    Max. 0.002 %
  • S-compounds (as S):
    Max. 0.003 %
  • APHA:
    Max. 10
  • Reaction against H₂SO₄:
    complying
  • Vapor Pressure:
    18 mmHg ( 37.7 °C)

 

 

Frequently Bought Together