ContactUsLinkComponent
Perchloric acid 0.1 M (0.1 N) in glacial acetic acid volumetric solution, for titration in non-aqueous media, Fluka™
Flammable
Corrosive
Perchloric acid 0.1 M (0.1 N) in glacial acetic acid volumetric solution, for titration in non-aqueous media, Fluka™
Certificates
undefined
Flammable
Corrosive
Perchloric acid 0.1 M (0.1 N) in glacial acetic acid volumetric solution, for titration in non-aqueous media, Fluka™
Specification Test Results
  • Assay:
    0.0997 - 0.1003 mol/l
  • Water (Karl Fischer):
    0.1 - 0.2 %
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective