Order Entry
Switzerland
ContactUsLinkComponent
Multi-element standards, 10 components in 2% HNO₃
Multi-element standards, 10 components in 2% HNO₃
Catalog # PERCN0691579
Supplier:  PerkinElmer
CAS Number:  
Multi-element standards, 10 components in 2% HNO₃
Catalog # PERCN0691579
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Description:
    Multi-element standard, 10 components
  • Pk Info:
    1× 500 ml

Specifications

About this item

1 mg/l of barium, magnesium

10 mg/l of lanthanum, lithium, manganese, nickel, strontium, zinc

50 mg/l of arsenic, potassium