Order Entry
Switzerland
ContactUsLinkComponent
Nitric acid 2%, Optima® for calibration blank in ICP
Corrosive
Nitric acid 2%, Optima® for calibration blank in ICP
Catalog # PERCN0773120
Supplier:  PerkinElmer
CAS Number:  
Corrosive
Nitric acid 2%, Optima® for calibration blank in ICP
Catalog # PERCN0773120
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    2730
  • Pk Info:
    1×500 ml

Specifications

About this item

 :
 :63.01 g/mol
UN:3264
ADR:8,III
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349