Calibration standard, single element, vanadium for AAS and ICP-OES, pure grade
Catalog # PERCN9303808
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pk:1
- Environmentally Preferable:
- Pk Info:1×125 ml
- Element:Vanadium
- Symbol:V
Specifications
About this item
Pure Grade Standards for AA and ICP-OES
- Analyzed by ICP-OES
- Analyzed by Classical Wet Assay
- 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppm level
- All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.